Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human YES1 (NP_005424.1, 1 a.a. - 543 a.a.) T348I mutant Full length Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5753

Catalog No. 89-967-599


Add to cart

Description

Description

This gene is the cellular homolog of the Yamaguchi sarcoma virus oncogene. The encoded protein has tyrosine kinase activity and belongs to the src family of proteins. This gene lies in close proximity to thymidylate synthase gene on chromosome 18, and a corresponding pseudogene has been found on chromosome 22. [provided by RefSeq]

Specifications

Specifications

Baculovirus infected insect cell (Sf21) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
88kDa
Baculovirus infected insect cell (Sf21) expression system
82% by SDS-PAGE/CBB staining
5 ug
RUO
YES1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Srctide. ATP: 100 µM.
Yes
NP_005424.1
Liquid
7525
YES1 (T348I) (Human) Recombinant Protein
Glutathione sepharose chromatography
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT441/P61-YES/Yes/c-yes
YES1
Insect
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit