Learn More
Raybiotech IncSupplier Diversity Partner Human YWHAE ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHYWHAE1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.34 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human YWHAE.
Catalog No. 50-194-9863