Learn More
Abnova™ Human ZADH1 Full-length ORF (NP_689657.1, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_689657.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 145482 |
Molecular Weight (g/mol) | 64.9kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-967-654
|
Abnova™
H00145482P01L |
25 ug |
Each for $769.00
|
|
|||||
89-967-653
|
Abnova™
H00145482P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes an enzyme involved in the metabolism of prostaglandins. The encoded protein catalyzes the NADPH-dependent conversion of 15-keto-prostaglandin E2 to 15-keto-13,14-dihydro-prostaglandin E2. This protein may also be involved in regulating activation of the peroxisome proliferator-activated receptor. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MIVQRVVLNSRPGKNGNPVAENFRMEEVYLPDNINEGQVQVRTLYLSVDPYMRCRMNEDTGTDYITPWQLSQVVDGGGIGIIEESKHTNLTKGDFVTSFYWPWQTKVILDGNSLEKVDPQLVDGHLSYFLGAIGMPGLTSLIGIQEKGHITAGSNKTMVVSGAAGACGSVAGQIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKEGKLKIKETVINGLENMGAAFQSMMTGGNIGKQIVCISEEISLSpecifications
NP_689657.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MIVQRVVLNSRPGKNGNPVAENFRMEEVYLPDNINEGQVQVRTLYLSVDPYMRCRMNEDTGTDYITPWQLSQVVDGGGIGIIEESKHTNLTKGDFVTSFYWPWQTKVILDGNSLEKVDPQLVDGHLSYFLGAIGMPGLTSLIGIQEKGHITAGSNKTMVVSGAAGACGSVAGQIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKEGKLKIKETVINGLENMGAAFQSMMTGGNIGKQIVCISEEISL | |
RUO | |
PTGR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
145482 | |
ZADH1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686P10120/FLJ39091/PGR-2/PGR2/PTRG2/ZADH1 | |
PTGR2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.