Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZBTB7B Partial ORF (NP_056956.1, 433 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ZBTB7B (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 37.29kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-967-710
View Documents
Abnova™
H00051043Q01L
25 ug
Each for $709.50
Add to cart
 
89-967-709
View Documents
Abnova™
H00051043Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.[supplied by OMIM]

Sequence: HLCHRAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAAFPAGLDLSNGHLDTFRLSLARFWEQSAPTWAPVSTPGPPDDDEEEGAPTTPQAEGAME
Specifications

Specifications

ZBTB7B (Human) Recombinant Protein (Q01)
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
37.29kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit