Learn More
Abnova™ Human ZFYVE1 Full-length ORF (NP_067083.1, 1 a.a. - 777 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_067083.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53349 |
Molecular Weight (g/mol) | 113.6kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-967-876
|
Abnova™
H00053349P01L |
25 ug |
Each for $769.00
|
|
|||||
89-967-875
|
Abnova™
H00053349P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes. [provided by RefSeq]
Sequence: MSAQTSPAEKGLNPGLMCQESYACSGTDEAIFECDECCSLQCLRCEEELHRQERLRNHERIRLKPGHVPYCDLCKGLSGHLPGVRQRAIVRCQTCKINLCLECQKRTHSGGNKRRHPVTVYNVSNLQESLEAEEMDEETKRKKMTEKVVSFLLVDENEEIQVTNEEDFIRKLDCKPDQHLKVVSIFGNTGDGKSHTLNHTFFYGREVFKTSPTQESCTVGVWAAYDPVHKVAVIDTEGLLGATVNLSQRTRLLLKVLAISDLVIYRTHADRLHNDLFKFLGDASEAYLKHFTKELKATTARCGLDVPLSTLGPAVIIFHETVHTQLLGSDHPSEVPEKLIQDRFRKLGRFPEAFSSIHYKGTRTYNPPTDFSGLRRALEQLLENNTTRSPRHPGVIFKALKALSDRFSGEIPDDQMAHSSFFPDEYFTCSSLCLSCGVGCKKSMNHGKEGVPHEAKSRCRYSHQYDNRVYTCKACYERGEEVSVVPKTSASTDSPWMGLAKYAWSGYVIECPNCGVVYRSRQYWFGNQDPVDTVVRTEIVHVWPGTDGFLKDNNNAAQRLLDGMNFMAQSVSELSLGPTKAVTSWLTDQIAPAYWRPNSQILSCNKCATSFKDNDTKHHCRACGEGFCDSCSSKTRPVPERGWGPAPVRVCDNCYEARNVQLAVTEAQVDDEGGTLIARKVGEAVQNTLGAVVTAIDIPLGLVKDAARPAYWVPDHEILHCHNCRKEFSIKLSKHHCRACGQGFCDECSHDRRAVPSRGWDHPVRVCFNCNKKPGDLSpecifications
NP_067083.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSAQTSPAEKGLNPGLMCQESYACSGTDEAIFECDECCSLQCLRCEEELHRQERLRNHERIRLKPGHVPYCDLCKGLSGHLPGVRQRAIVRCQTCKINLCLECQKRTHSGGNKRRHPVTVYNVSNLQESLEAEEMDEETKRKKMTEKVVSFLLVDENEEIQVTNEEDFIRKLDCKPDQHLKVVSIFGNTGDGKSHTLNHTFFYGREVFKTSPTQESCTVGVWAAYDPVHKVAVIDTEGLLGATVNLSQRTRLLLKVLAISDLVIYRTHADRLHNDLFKFLGDASEAYLKHFTKELKATTARCGLDVPLSTLGPAVIIFHETVHTQLLGSDHPSEVPEKLIQDRFRKLGRFPEAFSSIHYKGTRTYNPPTDFSGLRRALEQLLENNTTRSPRHPGVIFKALKALSDRFSGEIPDDQMAHSSFFPDEYFTCSSLCLSCGVGCKKSMNHGKEGVPHEAKSRCRYSHQYDNRVYTCKACYERGEEVSVVPKTSASTDSPWMGLAKYAWSGYVIECPNCGVVYRSRQYWFGNQDPVDTVVRTEIVHVWPGTDGFLKDNNNAAQRLLDGMNFMAQSVSELSLGPTKAVTSWLTDQIAPAYWRPNSQILSCNKCATSFKDNDTKHHCRACGEGFCDSCSSKTRPVPERGWGPAPVRVCDNCYEARNVQLAVTEAQVDDEGGTLIARKVGEAVQNTLGAVVTAIDIPLGLVKDAARPAYWVPDHEILHCHNCRKEFSIKLSKHHCRACGQGFCDECSHDRRAVPSRGWDHPVRVCFNCNKKPGDL | |
RUO | |
ZFYVE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53349 | |
ZFYVE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFCP1/KIAA1589/TAFF1/ZNFN2A1 | |
ZFYVE1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.