Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ZFYVE16 Full-length ORF (AAH30808.1, 1 a.a. - 809 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009765P01S
Description
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule (Seet et al., 2004 [PubMed 14613930]).[supplied by OMIM]
Sequence: MDSYFKAAVSDLDKLLDDFEQNPDEQDYLQDVQNAYDSNHCSVSSELASSQRTSLLPKDQECVNSCASSETSYGTNESSLNKKTLKGLTSIQNEKNVTGLDLLSSVDGGTSDEIQPLYMGRCSKPICDLISDMGNLVHATNSEEDIKKLLPDDFKSNADSLIGLDLSSVSDTPCVSSTDHDSDTVREQQNDTSSELQNREIGGIKELGIKVDTTLSDSYNYSGTENLKDKKIFNQLESIVDFNMSSALTRQSSKMFHAKDKLQHKSQPCGLLKDVGLVKEEVDVAVITAAECLKEEGKTSALTCSLPKNEDLCLNDSNSRDENFKLPDFSFQEDKTVIKQSAQEDSKSLDLKDNDVIQDSSSALHVSSKDVPSSLSCLPASGSMCGSLIESKARGDFLPQHEHKDNIQDAVTIHEEIQNSVVLGGEPFKENDLLKQEKCKSILLQSLIEGMEDRKIDPDQTVIRAESLDGGDTSSTVVESQEGLSGTHVPESSDCCEGFINTFSSNDMDGQDLDYFNIDEGAKSGPLISDAELDAFLTEQYLQTTNIKSFEENVNDSKSQMNQIDMKGLDDGNINNIYFNAEAGAIGESHGINIICETVDKQNTIENGLSLGEKSTIPVQQGLPTSKSEITNQLSVSDINSQSVGGARPKQLFSLPSRTRSSKDLNKPDVPDTIESEPSTADTVVPITCAIDSTADPQVSFNSNYIDIESNSEGGSSFVTANEDSVPENTCKEGLVLGQKQPTWVPDSEAPNCMNCQVKFTFTKRRHHCRACGKVFCGVCCNRKCKLQYLEKEARVCVVCYETISKGECSpecifications
AAH30808.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E13162/ENDOFIN/KIAA0305 | |
ZFYVE16 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9765 | |
ZFYVE16 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSYFKAAVSDLDKLLDDFEQNPDEQDYLQDVQNAYDSNHCSVSSELASSQRTSLLPKDQECVNSCASSETSYGTNESSLNKKTLKGLTSIQNEKNVTGLDLLSSVDGGTSDEIQPLYMGRCSKPICDLISDMGNLVHATNSEEDIKKLLPDDFKSNADSLIGLDLSSVSDTPCVSSTDHDSDTVREQQNDTSSELQNREIGGIKELGIKVDTTLSDSYNYSGTENLKDKKIFNQLESIVDFNMSSALTRQSSKMFHAKDKLQHKSQPCGLLKDVGLVKEEVDVAVITAAECLKEEGKTSALTCSLPKNEDLCLNDSNSRDENFKLPDFSFQEDKTVIKQSAQEDSKSLDLKDNDVIQDSSSALHVSSKDVPSSLSCLPASGSMCGSLIESKARGDFLPQHEHKDNIQDAVTIHEEIQNSVVLGGEPFKENDLLKQEKCKSILLQSLIEGMEDRKIDPDQTVIRAESLDGGDTSSTVVESQEGLSGTHVPESSDCCEGFINTFSSNDMDGQDLDYFNIDEGAKSGPLISDAELDAFLTEQYLQTTNIKSFEENVNDSKSQMNQIDMKGLDDGNINNIYFNAEAGAIGESHGINIICETVDKQNTIENGLSLGEKSTIPVQQGLPTSKSEITNQLSVSDINSQSVGGARPKQLFSLPSRTRSSKDLNKPDVPDTIESEPSTADTVVPITCAIDSTADPQVSFNSNYIDIESNSEGGSSFVTANEDSVPENTCKEGLVLGQKQPTWVPDSEAPNCMNCQVKFTFTKRRHHCRACGKVFCGVCCNRKCKLQYLEKEARVCVVCYETISKGEC | |
RUO | |
ZFYVE16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction