Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZIM2 Full-length ORF (NP_056178.3, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name ZIM2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 87.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-967-920
View Documents
Abnova™
H00023619P01L
25 ug
Each for $709.50
Add to cart
 
89-967-919
View Documents
Abnova™
H00023619P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5' exons and have a common promoter, and both genes are paternally expressed (PMID:15203203). Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Additional 5' alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene. [provided by RefSeq]

Sequence: MYQPEDDNNSDVTSDDDMTRNRRESSPPHSVHSFSGDRDWDRRGRSRDMEPRDRWSHTRNPRSRMPPRDLSLPVVAKTSFEMDREDDRDSRAYESRSQDAESYQNVVDLAEDRKPHNTIQDNMENYRKLLSLGFLAQDSVPAEKRNTEMLDNLPSAGSQFPDFKHLGTFLVFEELVTFEDVLVDFSPEELSSLSAAQRNLYREVMLENYRNLVSLGHQFSKPDIISRLEEEESYAMETDSRHTVICQGESHDDPLEPHQGNQEKLLTPITMNDPKTLTPERSYGSDEFERSSNLSKQSKDPLGKDPQEGTAPGICTSPQSASQENKHNRCEFCKRTFSTQVALRRHERIHTGKKPYECKQCAEAFYLMPHLNRHQKTHSGRKTSGCNEGRKPSVQCANLCERVRIHSQEDYFECFQCGKAFLQNVHLLQHLKAHEAARVLPPGLSHSKTYLIRYQRKHDYVGERACQCCDCGRVFSRNSYLIQHYRTHTQERPYQCQLCGKCFGRPSYLTQHYQLHSQEKTVECDHC
Specifications

Specifications

ZIM2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
87.6kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit