Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZMPSTE24 Partial ORF (NP_005848.2, 402 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010269Q01S

 View more versions of this product

Catalog No. 89-967-934


Add to cart

Description

Description

This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq]

Sequence: VLSFCLTVLSRRFEFQADAFAKKLGKAKDLYSALIKLNKDNLGFPVSDWLFSMWHYSHPPLLERLQALKTMKQH
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.88kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VLSFCLTVLSRRFEFQADAFAKKLGKAKDLYSALIKLNKDNLGFPVSDWLFSMWHYSHPPLLERLQALKTMKQH
RUO
ZMPSTE24
Wheat Germ (in vitro)
GST
NP_005848.2
Liquid
10269
ZMPSTE24 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FACE-1/FACE1/FLJ14968/STE24/Ste24p
ZMPSTE24
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit