Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF254 Partial ORF (NP_004867.2, 149 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009534Q02S

 View more versions of this product

Catalog No. 89-968-113


Add to cart

Description

Description

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]

Sequence: AQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKS
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.34kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKS
RUO
ZNF254
Wheat Germ (in vitro)
GST
NP_004867.2
Liquid
9534
ZNF254 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BMZF-5/FLJ58216/HD-ZNF1/ZNF539/ZNF91L
ZNF254
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit