Learn More
Abnova™ Human ZNF278 Full-length ORF (NP_114440.1, 1 a.a. - 537 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023598P01
Description
The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36.1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene. [provided by RefSeq]
Sequence: MERVNDASCGPSGCYTYQVSRHSTEMLHNLNQQRKNGGRFCDVLLRVGDESFPAHRAVLAACSEYFESVFSAQLGDGGAADGGPADVGGATAAPGGGAGGSRELEMHTISSKVFGDILDFAYTSRIVVRLESFPELMTAAKFLLMRSVIEICQEVIKQSNVQILVPPARADIMLFRPPGTSDLGFPLDMTNGAALAANSNGIAGSMQPEEEAARAAGAAIAGQASLPVLPGVDRLPMVAGPLSPQLLTSPFPSVASSAPPLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACEICGKIFRDVYHLNRHKLSHSGEKPYSCPVCGLRFKRKDRMSYHVRSHDGSVGKPYICQSCGKGFSRPDHLNGHIKQVHTSERPHKCQVWVGSSSGLPPLEPLPSDLPSWDFAQPALWRSSHSVPDTAFSLSLKKSFPALENLGPAHSSNTLFCPAPPGYLRQGWTTPEGSRAFTQWPVGSpecifications
NP_114440.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAZR/PATZ/RIAZ/ZBTB19/ZNF278/ZSG/dJ400N23 | |
PATZ1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23598 | |
ZNF278 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERVNDASCGPSGCYTYQVSRHSTEMLHNLNQQRKNGGRFCDVLLRVGDESFPAHRAVLAACSEYFESVFSAQLGDGGAADGGPADVGGATAAPGGGAGGSRELEMHTISSKVFGDILDFAYTSRIVVRLESFPELMTAAKFLLMRSVIEICQEVIKQSNVQILVPPARADIMLFRPPGTSDLGFPLDMTNGAALAANSNGIAGSMQPEEEAARAAGAAIAGQASLPVLPGVDRLPMVAGPLSPQLLTSPFPSVASSAPPLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACEICGKIFRDVYHLNRHKLSHSGEKPYSCPVCGLRFKRKDRMSYHVRSHDGSVGKPYICQSCGKGFSRPDHLNGHIKQVHTSERPHKCQVWVGSSSGLPPLEPLPSDLPSWDFAQPALWRSSHSVPDTAFSLSLKKSFPALENLGPAHSSNTLFCPAPPGYLRQGWTTPEGSRAFTQWPVG | |
RUO | |
PATZ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.