Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNFN1A1 Full-length ORF (AAH18349.1, 1 a.a. - 477 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010320P01L

 View more versions of this product

Catalog No. 89-948-583

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Sequence: MDADEGQDMSQVSGKESPPVSDTPDEGDEPMPIPEDLSTTSGGQQSSKSDRVVASNVKVETQSDEENGRACEMNGEECAEDLRMLDASGEKMNGSHRDQGSSALSGVGGIRLPNGKLKCDICGIICIGPNVLMVHKRSHTGERPFQCNQCGASFTQKGNLLRHIKLHSGEKPFKCHLCNYACRRRDALTGHLRTHSVIKEETNHSEMAEDLCKIGSERSLVLDRLASNVAKRKSSMPQKFLGDKGLSDTPYDSSASYEKENEMMKSHVMDQAINNAINYLGAESLRPLVQTPPGGSEVVPVISPMYQLHKPLAEGTPRSNHSAQDSAVENLLLLSKAKLVPSEREASPSNSCQDSTDTESNNEEQRSGLIYLTNHIAPHARNGLSLKEEHRAYDLLRAASENSQDALRVVSTSGEQMKVYKCEHCRVLFLDHVMYTIHMGCHGFRDPFECNMCGYHSQDRYEFSSHITRGEHRFHMS
Specifications

Specifications

AAH18349.1
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.1kDa
Glutathione Sepharose 4 Fast Flow
25μg
RUO
IKZF1
Wheat Germ (in vitro)
GST Tag
wheat germ expression system
Liquid
10320
Human ZNFN1A1 Full-length ORF (AAH18349.1, 1 a.a. - 477 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Hs.54452, IK1, IKAROS, LYF1, PRO0758, ZNFN1A1, hIk-1
IKZF1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit