Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone Rabbit anti-Human, Clone: JM108-09, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266893
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Growth Hormone | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-400 | |
GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
Rabbit | |
Protein A purified | |
Biologically Active Proteins | |
2688 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
JM108-09 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
GH1 | |
Synthetic peptide within Human Growth Hormone aa 72-106/ 217. (SwissProt: P01241 Human) | |
100 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction