Learn More
Novus Biologicals Human MTR ELISA Kit (Colorimetric)
Human MTR ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer: Novus Biologicals NBP276709
Description
The Human MTR ELISA Kit (Colorimetric) measures human MTR in serum, plasma and other biological fluids.Specifications
0.781 to 50 ng/mL | |
ELISA Kit (Colorimetric) | |
Storage is content dependent. | |
MTR | |
Biological Fluids, Plasma, Serum |
4548 | |
0.469 ng/mL | |
Human | |
1 Kit |