Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HumanSerumAlbumin Mouse anti-Human, Clone: 8-A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266895

Catalog No. NBP266895


Add to cart

Description

Description

HumanSerumAlbumin Monoclonal antibody specifically detects HumanSerumAlbumin in Human samples. It is validated for Western Blot.
Specifications

Specifications

HumanSerumAlbumin
8-A3
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Mouse
IgG1
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot
2 mg/mL
Western Blot 1:2000
Purified
ALB
Protein
Protein A purified
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary
213
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit