Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HumanSerumAlbumin Rabbit anti-Bovine, Clone: JF32-10, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266894
Description
Human Serum Albumin Monoclonal specifically detects Human Serum Albumin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HumanSerumAlbumin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Rabbit | |
Protein A purified | |
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JF32-10 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
ALB | |
Synthetic peptide within Human Human Serum Albumin aa 156-189/ 609. (SwissProt: P02768 Human; SwissProt: P07724 Mouse; SwissProt: P02770 Rat; SwissProt: P02769 Cow) | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction