Learn More
Humboldt Filter Pump Aspirators
Offer excellent performance for vacuum filtrations under both high and low water pressure conditions
Manufacturer:  Humboldt H12090
 View more versions of this product
Catalog No. 09-790-24A
Add to cart
Description
Description
  • Swirling type, available in two models: Chapman and Short
  • Nickel-plated
  • Ball-type, positive-action valve in suction tube prevents water backflow if pressure is reduced
  • Water inlet: 3/8 in., NPT tapered
Specifications
Specifications
Zinc - Nickel-plated
76mm
3 in.