Learn More
Humboldt Micro Burner
Designed for micro-methods of analysis
Manufacturer:  Humboldt H5815
 View more versions of this product
Catalog No. 22-305636
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Providing a small, intense flame, these burners are perfect for micro-methods of analysis.
  • Die-cast, nickel-plated zinc alloy base
  • Serrated gas inlet connections
  • Aluminum mixing tube
  • Overall height: 3.5 in. (8.75cm)