Learn More
Mercodia Inc HUMN ULTR SENSITIV C-PEPT ELIS
SDP
Supplier:  Mercodia Inc 10114101
Encompass_Preferred
Immunoassay for sensitive and specific quantitative determination of human C-peptide in urine, serum or plasma. Offers a high sensitivity detection limit of <2.5 pmol/L. No cross reactivity to insulin or proinsulin.
Catalog No. NC0125912
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.