Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Huntingtin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302813A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Huntington′s disease (HD) is a neurodegenerative disorder caused by an expanding polyglutamine repeat in the huntingtin gene. HD is a mid-life onset autosomal domit neurodegenerative disease that is characterized by psychiatric disorders, dementia, and involuntary movements (chorea), leading to death in 10-20 years. The HD gene product is widely expressed in human tissues, with the highest level of expression in the brain. The huntington gene product is expressed at similar levels in patients and controls, which suggests that the expansion of the polyglutamine repeat induces a toxic gain of function perhaps through interactions with other cellular proteins. Using yeast two-hybrid system Li et al., have identified a protein, HAP1 (huntington associated protein 1), that associates with huntingtin protein. The in vitro data suggest that the association between HAP1 and huntington is enhanced by increasing length of glutamine repeat.Specifications
Huntingtin | |
Polyclonal | |
Unconjugated | |
HTT | |
HD, IT15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3064 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P42858 | |
HTT | |
Between 2300 and 2350 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title