Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Huntingtin Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 199571AP

Catalog No. 50-173-2623


Add to cart

Description

Description

The antibody recognizes the C-term of HTT.

This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications

Specifications

Huntingtin
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P42858, P42859
HD, HD protein, HTT, huntingtin, Huntington disease protein, IT15
Rabbit
IgG
150 μL
-20°C
Primary
15194, 3064
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
HTT
Liquid
Htt
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit