Learn More
Selleck Chemical LLCSupplier Diversity Partner (-)-Huperzine A (HupA) 5mg 102518-79-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S22515MG
(-)-Huperzine A is a potent, highly specific and reversible inhibitor of acetylcholinesterase (AChE) with Ki of 7 nM, exhibiting 200-fold more selectivity for G4 AChE over G1 AChE. Also acts as an NMDA receptor antagonist. Phase 4.
Catalog No. 50-706-3