Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuR/ELAVL1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI14F4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270635AF647

Catalog No. N270635A647


Add to cart

Description

Description

HuR/ELAVL1 Monoclonal antibody specifically detects HuR/ELAVL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

HuR/ELAVL1
OTI14F4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ELAVL1
Full length recombinant protein of human HuR/ELAVL1 (NP_001410) produced in HEK293T cells.
Immunogen affinity purified
Vision
Primary
1994.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit