Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HuR/ELAVL1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30294720UL

 View more versions of this product

Catalog No. NB092382


Add to cart

Description

Description

HuR/ELAVL1 Polyclonal antibody specifically detects HuR/ELAVL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Specifications

Specifications

HuR/ELAVL1
Unconjugated
Purified
Rabbit
IgG
20 μL
Vision
Primary
1994
Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot
PBS with 50% glycerol, pH7.3.
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HuR/ ELAVL1 (NP_001410.2).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit