Learn More
Aviva Systems Biology Corp HUS1B Antibody - N-terminal region (ARP52871_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HUS1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HUS1B. Alternate names: MGC126746, MGC126748, RP11-532F6.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60858