Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HUWE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520374
Description
A suggested positive control is Daudi cell lysate. PA5-20374 can be used with blocking peptide PEP-0491.
SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
Specifications
HUWE1 | |
Polyclonal | |
Unconjugated | |
HUWE1 | |
5430439H10Rik; ARF-binding protein 1; ARF-BP1; AU041296; BJ-HCC-24 tumor antigen; C430014N20Rik; C80292; E3 ubiquitin-protein ligase HUWE1; E3Histone; Gm1718; HECT domain protein LASU1; HECT, UBA and WWE domain containing 1; HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase; HECT, UBA and WWE domain-containing protein 1; HectH9; HECT-type E3 ubiquitin transferase HUWE1; Homologous to E6AP carboxyl terminus homologous protein 9; HSPC272; HUWE1; Ib772; Kiaa0312; KIAA1578; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; RP3-339A18.4; upstream regulatory element binding protein 1; upstream regulatory element-binding protein 1; Ureb1; URE-B1; URE-binding protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10075, 501546, 59026 | |
4°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P51593, Q7TMY8, Q7Z6Z7 | |
HUWE1 | |
A 19 amino acid peptide from near the carboxy terminus of human ARF-BP1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction