Learn More
HUWE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 194301AP
Catalog No. 50-173-2403
Add to cart
Description
Description
SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
Specifications
Specifications
HUWE1
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51593, Q7TMY8, Q7Z6Z7
ARF binding protein 1, ARF BP1, HECTH9, HSPC272, HUWE1, Ib772, KIAA0312, KIAA1578, Large structure of UREB1, LASU1, Mcl 1 ubiquitin ligase E3, MULE, URE B1, URE binding protein 1, UREB1
Rabbit
IgG
150 μL
-20°C
Primary
10075, 501546, 59026
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HUWE1
Liquid
Huwe1
HUWE1 Fusion Protein Ag13763
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat