Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner Hy-Pip Cova PIP Specificity Plate  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner H63001EA
Hy-Pip Cova PIP Specificity plates are 96-well clear polystyrene microplate where each row has an individual phosphoinositide coated at 20 pmols per well and blocked. Hy-Pip Cova PIP Specificity plates are useful for determining the specificity of proteins against all eight phosphoinositides.
Catalog No. 50-720-376
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.