Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550426

Catalog No. PA550426


Add to cart

Description

Description

The antibody detects endogenous levels of total HYAL2 protein.

This gene encodes a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. Hyaluro is thought to be involved in cell proliferation, migration and differentiation. Varying functions have been described for this protein. It has been described as a lysosomal hyaluronidase which is active at a pH below 4 and specifically hydrolyzes high molecular weight hyaluro. It has also been described as a GPI-anchored cell surface protein which does not display hyaluronidase activity but does serve as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5 ft. UTR.
Specifications

Specifications

HYAL2
Polyclonal
Unconjugated
HYAL2
Liquid
HYAL2
Fusion protein corresponding to a region derived from internal residues of human hyaluronoglucosaminidase 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q12891
LUCA2
Rabbit
IgG
100 μL
-20°C
Primary
8692
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit