Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$204.00 - $482.50

Specifications

Antigen HYAL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094569
SDP
View Documents
Novus Biologicals
NBP30499520UL
20 μL
Each for $204.00
Add to Cart
 
NB094568
SDP
View Documents
Novus Biologicals
NBP304995100UL
100 μL
Each for $482.50
Add to Cart
 
Description

Description

HYAL2 Polyclonal specifically detects HYAL2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

HYAL2
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
8692
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
EC 3.2.1.35, Hyal-2, hyaluronidase 2, hyaluronoglucosaminidase 2, hyaluronoglucosaminidase-2, LuCa-2, LUCA2hyaluronidase-2, Lung carcinoma protein 2, lysosomal hyaluronidase, PH20 homolog, PH-20 homolog
Recombinant fusion protein containing a sequence corresponding to amino acids 367-473 of human HYAL2 (NP_149348.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit