Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HYAL3 Recombinant Protein

Manufacturer:  Abnova™ H00008372P0110

 View more versions of this product

Catalog No. 89-011-239

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH05896
Solution
8372
HYAL3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HYAL3
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
71.6
8
Glutathione Sepharose 4 Fast Flow
10 ug
MTTQLGPALVLGVALCLGCGQPLPQVPERPFSVLWNVPSAHCEARFGVHLPLNSLGIIANRGQHFHGQNMTIFYKNQLGLYPYFGPRGTAHNGGIPQALPLDRHLALAAYQIHHSLRPGFAGPAVLDWEEWCPLWAGNWGRRRAYQAASWAWAQQVFPDLDPQEQLYKAYTGFEQAARALMEDTLRVAQALRPHGLWGFYHYPACGNGWHSMASNYTGRCHAATLARNTQLHWLWAASSALFPSIYLPPRLPPAHHQAFVRHRLEEAFRVALVGHRHPLPVLAYVRLTHRRSGRFLSQDDLVQSIGVSAALGAAGVVLWGDLSLSSSEEECWHLHDYLVDTLGPYVINVTRAAMACSHQRCHGHGRCARRDPGQMEAFLHLWPDGSLGDWKSFSCHCYWGWAGPTCQEPRPGPKEAV
LUCA-3/LUCA14/LUCA3/Minna14
HYAL3
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit