Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ HYAL3 Recombinant Protein
Manufacturer: Abnova™ H00008372P0125
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH05896 | |
Solution | |
8372 | |
HYAL3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HYAL3 | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
71.6 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MTTQLGPALVLGVALCLGCGQPLPQVPERPFSVLWNVPSAHCEARFGVHLPLNSLGIIANRGQHFHGQNMTIFYKNQLGLYPYFGPRGTAHNGGIPQALPLDRHLALAAYQIHHSLRPGFAGPAVLDWEEWCPLWAGNWGRRRAYQAASWAWAQQVFPDLDPQEQLYKAYTGFEQAARALMEDTLRVAQALRPHGLWGFYHYPACGNGWHSMASNYTGRCHAATLARNTQLHWLWAASSALFPSIYLPPRLPPAHHQAFVRHRLEEAFRVALVGHRHPLPVLAYVRLTHRRSGRFLSQDDLVQSIGVSAALGAAGVVLWGDLSLSSSEEECWHLHDYLVDTLGPYVINVTRAAMACSHQRCHGHGRCARRDPGQMEAFLHLWPDGSLGDWKSFSCHCYWGWAGPTCQEPRPGPKEAV | |
LUCA-3/LUCA14/LUCA3/Minna14 | |
HYAL3 | |
Wheat Germ (in vitro) | |
GST |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title