Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548622

Catalog No. PA548622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Endo-hyaluronidase that degrades hyaluro to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.
Specifications

Specifications

HYAL4
Polyclonal
PBS with 0.09% sodium azide
Q2M3T9
Hyaluronidase-4, Hyal-4, Chondroitin sulfate endo-beta-N-acetylgalactosaminidase, Chondroitin sulfate hydrolase, CSHY, Hyaluronoglucosaminidase-4, HYAL4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23553
Western Blot
Unconjugated
HYAL4
Liquid
HYAL4
KLH conjugated synthetic peptide between 143-172 amino acids from the Central region of human HYAL4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit