Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronan synthase 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Hyaluronan synthase 1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094874
View Documents
Novus Biologicals
NBP303814100UL
100 μg
Each for $320.71
Add to cart
 
NB094875
View Documents
Novus Biologicals
NBP30381420UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Hyaluronan synthase 1 Polyclonal antibody specifically detects Hyaluronan synthase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Hyaluronan synthase 1
Unconjugated
Rabbit
Human, Mouse
HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1
Recombinant fusion protein containing a sequence corresponding to amino acids 80-390 of human Hyaluronan synthase 1 (NP_001514.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
3036
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit