Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Hyaluronic acid sodium salt, Streptococcus equi, 91%

Click to view available options
Quantity:
500mg
1g
9067-32-7
MFCD01773053
9067-32-7
MFCD01773053
Applications
Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Molecular weight is greater than or equal to 1MDa
Merck Index 14,4757
Solubility Information Soluble in water
Physical Form Powder
Percent Purity 91%
Color White
Quantity 500mg
Chemical Name or Material Hyaluronic acid sodium salt, Streptococcus equi

RTECSNumber : MT7250000

storageNote1 : -30° to -10°C