Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyamine Hydroxide, Fisher BioReagents

Click to view available options
Quantity:
500mL
Packaging:
Amber Glass
26248-39-5
C28H45NO3
443.672
MJAMCTLGWXIKOT-UHFFFAOYSA-M
hyamine hydroxide, methylbenzethonium hydroxide, benzyldimethyl 2-2-4-1,1,3,3-tetramethylbutyl tolyloxy ethoxy ethyl ammonium hydroxide, benzenemethanaminium, n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-, hydroxide, n-benzyl-n,n-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethanaminium hydroxide, benzenemethanaminium,n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-,hydroxide 9ci, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide
168497
benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;hydroxide
CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[OH-]
26248-39-5
C28H45NO3
443.672
MJAMCTLGWXIKOT-UHFFFAOYSA-M
hyamine hydroxide, methylbenzethonium hydroxide, benzyldimethyl 2-2-4-1,1,3,3-tetramethylbutyl tolyloxy ethoxy ethyl ammonium hydroxide, benzenemethanaminium, n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-, hydroxide, n-benzyl-n,n-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethanaminium hydroxide, benzenemethanaminium,n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-,hydroxide 9ci, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide
168497
benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;hydroxide
CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[OH-]
Melting Point -30°C
Packaging Amber Glass
Color Colorless
Identification Pass Test
Boiling Point 65°C
Density 0.933g/cm³
Quantity 500mL
Formula Weight 443.7
Physical Form Liquid
Chemical Name or Material Hyamine Hydroxide™

WARNING!

Emergency Overview
DANGER!. Keep away from open flames, hot surfaces and sources of ignition. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:3
Instability:1

Recommended Storage : RT