Learn More
Quality Biological IncSupplier Diversity Partner Hybridization Buffer, 500 mL DFS Item
Manufacturer:  Quality Biological IncSupplier Diversity Partner 351078101
A hybridization buffer specifically formulated for use with nylon membranes. • It does not contain Denhardt’s Reagent, which may interfere with the hybridization reaction of DNA immobilized on nylon. • Formulated for Nylon membrane • Filtered through a 0.22 µm membrane • DNase, RNase, and Protease tested • Applications: Denatured fragmented salmon sperm DNA may be added to this hybridization buffer to reduce nonspecific background. • Storage Conditions: 25°C • Shipping Condition: ambient temperature • Sterile Filtered • DNase Tested • Product and/or packaging contains recycled/renewable content • Product and/or packaging is recyclable or reduces waste • Product and/or packaging is safer to human and environmental health • Harmonization Code: 3822.00.5090 • ECCN: EAR99 • Max Shelf Life: 36 months
Catalog No. 50-146-835