Learn More
MilliporeSigma™ HybridSPE™-Phospholipid 96-well Plate
HybridSPE™ Phospholipid Removal Technology for Biological Matrices. Remove phospholipids and proteins for accurate and reproducible LC-MS analysis.
Supplier: MilliporeSigma™ 575657U
Description
Key Features and Benefits:
- More accurate results
- Unique chemistry can effectively separate hydrophobic analytes (such as vitamins) from phospholipids, unlike competitive product chemistries
- Elimination of proteins and phospholipid-induced ion suppression
- Simultaneous removal of proteins and phospholipids
- Simple, standardized methodology, analogous to traditional protein precipitation
- Alternative to complex traditional SPE method development
- Reproducible, consistent performance reduces need for reprocessing
- Less instrument downtime and longer column life
- Decreased run times by eliminating the need for gradients to clean columns between samples
- High throughput processing that is automatable and compatible with common robotic systems
- Ready-to-use, no preconditioning required
Specifications
50 mg | |
Fritless SPE Cartridge and Plates | |
20/Pk. |
Food and Beverage | |
2 mL |