Learn More
Cytiva HyCell™ TransFx™-C Media
For transient transfection of CHO cells and production of a variety of recombinant proteins manufactured under cGMP in ISO-compliant facilities
$125.60
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SH3094102
View Documents Promotion Details
Cytiva
SH30941.02
1 L bottle
Each for $125.60
Add to cart
Description
Description
  • Defined hydrolosate-free ADCF cell culture media formulation
  • Supports transient transfection and growth of multiple CHO cell lines
  • Supports the transient expression of a variety of recombinant proteins
  • Same media for transient transfection and production
  • Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
  • Component traceability
  • Manufactured under cGMP in ISO-compliant facility
Order Info
Available in liquid form in 1L bottles, 5 to 500L bioprocess containers or custom packaging. The media is also available as a powder in pack sizes from 5 to 1,000L for bioprocess production applications.