Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
GE Healthcare HyClone™ AdvanceSTEM Serum Replacement
Supports the growth of undifferentiated pluripotent embryonic stem cells.
Manufacturer:  GE Healthcare SH30874.02
Catalog No. SH3087402
Description
Description
  • Designed to maintain mESCs in an undifferentiated state
  • Intended for use with HyClone AdvanceSTEM Low Osmo DMEM
  • Serum-free formulation
  • Specifications
    Specifications
    Embryonic Stem Cells (ESCs) and Induced Pluripotent Stem Cells (iPSCs) 
    100 mL
    Store frozen
    Liquid
    100 mL
    Without: Serum; Leukemia Inhibitory Factory (LIF) 