Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ Antibiotic/Antimycotic Solution (Pen/Strep/Fungiezone) solution

Filter sterilized. GE Healthcare HyClone™ Antibiotic Antimycotic (Pen/Strep/Fungiezone) Solution controls or destroys destructive microorganisms such as bacteria, fungi and yeast.

Manufacturer:  GE Healthcare Life Sciences SV30079.01

Catalog No. SV3007901


Add to cart

Description

Description

  • Contains 10,000 units of penicillin, 10,000µg of streptomycin, and 25µg of Amphotericin B per mL
  • Wide spectrum of activity against gram negative and gram positive bacteria as well as activity against fungi and yeast
  • Commonly used as an antifungal, anti-yeast, anti-mold or antibacterial
  • Common working dose is 10mL/L of 100X solution
Specifications

Specifications

Broad spectrum antibacterial and antifungal
Antibiotic Antimycotic Solution (Pen/Strep/Fungizone), 100X
100 mL
Store frozen
100X
Liquid
0.2 μm sterile filtered
Safety and Handling

Safety and Handling

ShelfLife : 18 months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit