Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Calf Serum (U.S.) PROMO

A cost-effective alternative to Fetal Bovine Serum (FBS).

$84.30 - $108.50

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Sterilization Method Filtered through sequential 100 nm pore-size rated filters.
Storage Store at ≤-10°C
USDA Safety Testing Included Yes
Sterility Sterile
View More Specs

Products 2
Catalog Number Mfr. No. Heat Inactivated Price Quantity & Availability  
Catalog Number Mfr. No. Heat Inactivated Price Quantity & Availability  
SH300730LR1
View Documents
Cytiva
29648034
No
Each for $84.30
Add to cart
 
SH7304HILR1
View Documents
Cytiva
29648035
Yes
Each for $108.50
Add to cart
 
Description

Description

HyClone Bovine Calf Serum is available with or without iron supplementation. Bovine calf serum products contain exceptionally high levels of transferrin, which, when supplemented, can provide three to four times as much available iron as FBS. In many applications, the performance of calf serum will equal or even surpass that of FBS. Bovine Calf Serum is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

Specifications

Specifications

Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Store at ≤-10°C
Sterile
Filtered through sequential 100 nm pore-size rated filters.
Yes
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit