Learn More
HyClone™ CDM4CHO™ Media, Liquid: PROMO
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
Manufacturer:  GE Healthcare Life Sciences SH30558.02PM
 View more versions of this product
Catalog No. SH-305-5802LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
  • Format: Liquid
Specifications
Specifications
cGMP
Liquid
2° to 8°C
L-glutamine, phenol red, HEPES, protein
CDM4CHO media
1000mL
Sodium bicarbonate, poloxamer 188