Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ CDM4CHO™ Media, Liquid: PROMO

Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.

Specifications

Certifications/Compliance cGMP
Format Liquid
Storage Requirements 2° to 8°C
Description CDM4CHO media
View More Specs

Products 2
Catalog Number Mfr. No. Quantity With Additives Without Additives Price Quantity & Availability  
Catalog Number Mfr. No. Quantity With Additives Without Additives Price Quantity & Availability  
SH-305-5702LR View Documents GE Healthcare Life Sciences
SH30557.02PM
1000mL L-glutamine, sodium bicarbonate, poloxamer 188 Animal Defined Component, Protein, Phenol Red, HEPES This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
SH-305-5802LR View Documents GE Healthcare Life Sciences
SH30558.02PM
1000mL Sodium bicarbonate, poloxamer 188 L-glutamine, phenol red, HEPES, protein This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Format: Liquid
Specifications

Specifications

cGMP
Liquid
2° to 8°C
CDM4CHO media
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit