Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

GE Healthcare HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)

Enhance growth of cells with New Zealand sourced Characterized FBS.

Manufacturer:  GE Healthcare SH30406.01

 View more versions of this product

Catalog No. SH3040601



Description

Description

  • Carefully collected, processed, and filtered in our own New Zealand facility
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • Greater security against bovine disease, since New Zealand has the fewest reported bovine diseases in the world
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles using true pool technology
  • Specifications

    Specifications

    Sterile
    Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen
    ≤ 25 mg/dL
    None
    100 mL
    Yes
    Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus
    USDA Approved
    ≤ 20 EU/mL
    Yes
    Filtered through sequential 100 nm pore-size rated filters.
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Characterized FBS New Zealand origin
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit