Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HyClone™ Cosmic Calf Serum (New Zealand origin)

A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.

Manufacturer:  GE Healthcare SH30413.03

 View more versions of this product

Catalog No. SH3041303


Add to cart

Description

Description

All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability.Cosmic Calf Serum, New Zealand Origin is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

Specifications

Specifications

Sterile
≤ 25 EU/mL
Yes
Filtered through sequential 100 nm pore-size rated filters.
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Cosmic Calf Serum, New Zealand origin
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
≤ 25 mg/dL
None
1 L
Yes
Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit