Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Hyclone™ Donor Equine Serum
Carefully collected from selected U.S. donor herds that receive regular veterinary inspection and care.
$162.00
Specifications
Additional Assays | Albumin |
---|---|
Country of Origin | United States |
Product Type | Equine Serum |
Quantity | 1 L |
Sterility | Sterile |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
SH3007404
|
Cytiva
SH30074.04 |
1 L | None |
Each of 1 for $162.00
|
|
Description
Serum Collection:- From collection to final packaging
- Every step is documented
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Aseptically packaged in a Class 100 clean room condition
- Each lot of serum is tested to ensure quality and sterility
- Pooled after filtration and before final packaging
- Consistent quality from the first bottle filled to the last
- Increases consistency and minimizes variability with a lot of serum
Specifications
Albumin | |
Equine Serum | |
Sterile | |
No |
United States | |
1 L | |
Sequential Triple Filtered, 100 nm | |
HyClone™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title