Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ Fetal Bovine Serum (Canada), Characterized PROMO

Enhance cell culture growth with Canada sourced FBS tested for endotoxin, hemoglobin, and additional biochemical assays.

Manufacturer:  GE Healthcare Life Sciences 29334880

 View more versions of this product

Catalog No. SH33963HIPM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EMEA Certified
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Choride
Yes
≤25mg/dL
No
<-1°C
500mL
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV
Characterized Fetal Bovine Serum, Canada Origin
≤25EU/mL
Characterized
Canada
Heat Inactivated
Filtered through three sequential 100nm pore-size rated filters.
USDA Inspected
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit