Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ Fetal Bovine Serum (Canada), Characterized PROMO

Enhance cell culture growth with Canada sourced FBS tested for endotoxin, hemoglobin, and additional biochemical assays.

Specifications

Additional Assays Albumin
Certifications/Compliance EMEA Certified
Description Characterized Fetal Bovine Serum, Canada Origin
Endotoxin Concentration ≤25EU/mL
Grade Characterized
View More Specs

Products 2
Catalog Number Mfr. No. Heat Inactivated Processing Price Quantity & Availability  
Catalog Number Mfr. No. Heat Inactivated Processing Price Quantity & Availability  
SH3039603PM
View Documents
Cytiva - formerly GE Healthcare Life Sciences
29361669
No None This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
SH33963HIPM
View Documents
Cytiva - formerly GE Healthcare Life Sciences
29334880
Yes Heat Inactivated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

Albumin
Characterized Fetal Bovine Serum, Canada Origin
Characterized
No
500mL
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV
EMEA Certified
≤25EU/mL
≤25mg/dL
Canada
Filtered through three sequential 100nm pore-size rated filters.
USDA Inspected
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit