Learn More
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Fetal Bovine Serum (U.S.), Standard
Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) SH30088.03IR
 View more versions of this product
Catalog No. SH3008803IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Support cell culture growth
  • Standard grade
  • Aseptically packaged in Class 100 cleanroom
Specifications
Specifications
Sterile
Standard Fetal Bovine Serum, US Origin
For Information Only
Standard
U.S.
Gamma Irradiated
Filtered through sequential 100 nm pore-size rated filters.
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Standard Fetal Bovine Serum
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG
No
For Information Only
No
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
500mL
USDA Inspected
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Documents
Accessories