Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Clone I Serum PROMO

HyClone™ FetalClone™ I is a fetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.

$177.75

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration ≤ 20 mg/dL
IgG levels Typically less than 2 μg/mL
View More Specs

Products 1
Catalog Number Mfr. No. Heat Inactivated Price Quantity & Availability  
Catalog Number Mfr. No. Heat Inactivated Price Quantity & Availability  
SH3008HILR1
View Documents
Cytiva
29648028
Yes
Each for $177.75
Add to cart
 
Description

Description

Specifications

Specifications

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
≤ 10 EU/mL
Typically less than 2 μg/mL
No
Store at –10°C or below
U.S.
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
≤ 20 mg/dL
U.S.
Filtered using sequential 100 nm pore-size rated filters.
USDA Inspected
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit